Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Chloride, 11.8% (w/v) Solution, ASTM, For Sulfate, Spectrum™

Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
Grade Chemical Solution
CAS 10326-27-9,7732-18-5
Packaging Poly Bottle
Quantity 1 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.