Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Chloride, 12% (w/w), Reagents

Catalog No. 01335807
Change view
Click to view available options
Quantity:
20 L
Packaging:
Cubitainer
Hedpak
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-335-807 20 L Hedpak
01-335-806 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-807 Supplier Reagents Holdings Llc Supplier No. CB10260020E3
Only null left
Add to Cart
Add to Cart

Barium Chloride, 12% (w/w), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 10326-27-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Boiling Point 101°C
Color Colorless
Physical Form Liquid
Quantity 20 L
Percent Purity 12%
Grade Reagent
Packaging Hedpak
Specific Gravity 1.1148
Chemical Name or Material Barium Chloride, 12% (w/w)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.