Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Chloride, 20% (w/v) Solution, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Poly Bottle
Poly-Coated Glass Bottle
10326-27-9,7732-18-5
10326-27-9,7732-18-5
Grade Chemical Solution
Percent Purity 20% ± 0.5%
Packaging Poly-Coated Glass Bottle
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.