Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Chloride, Anhydrous, Powder, Purified, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
10361-37-2
BaCl2
208.227
WDIHJSXYQDMJHN-UHFFFAOYSA-L
barium chloride,barium dichloride,barium chloride,barium chloride solution,ccris 2286,barium chloride, anhydrous,barium chloride, ultra dry,bariumchlorid,dichlorobarium,dsstox_cid_24508
25204
CHEBI:63317
barium(2+);dichloride
[Cl-].[Cl-].[Ba+2]
10361-37-2
BaCl2
208.227
WDIHJSXYQDMJHN-UHFFFAOYSA-L
barium chloride,barium dichloride,barium chloride,barium chloride solution,ccris 2286,barium chloride, anhydrous,barium chloride, ultra dry,bariumchlorid,dichlorobarium,dsstox_cid_24508
25204
CHEBI:63317
barium(2+);dichloride
[Cl-].[Cl-].[Ba+2]
Formula Weight 208.23
Physical Form Powder
Grade Purified
Packaging Poly Bottle
Specific Gravity 1L = 3.86kg
Quantity 500g
Chemical Name or Material Barium Chloride, Anhydrous