Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium chloride dihydrate, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
500g
1kg
2.5kg
Packaging:
Plastic bottle
10326-27-9
BaCl2·2H2O
244.28
MFCD00149154
PWHCIQQGOQTFAE-UHFFFAOYSA-L
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
5284346
CHEBI:86153
barium(2+);dichloride;dihydrate
O.O.[Cl-].[Cl-].[Ba+2]
10326-27-9
BaCl2·2H2O
244.28
MFCD00149154
PWHCIQQGOQTFAE-UHFFFAOYSA-L
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
5284346
CHEBI:86153
barium(2+);dichloride;dihydrate
O.O.[Cl-].[Cl-].[Ba+2]
For analysis
Assay Percent Range 99% min. (Argentometry)
Linear Formula BaCl2·2H2O
Merck Index 15, 967
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,practically insoluble in ethanol,acetone and,ethyl acetate
Formula Weight 244.28
Physical Form Metal
Percent Purity 99+%
Grade Analysis
Calcium (Ca) 500ppm max.
Heavy Metals (as Pb) 5ppm max.
Iron (Fe) 2ppm max.
Loss on Drying 14.0 % to 16.0 % (150°C)
Packaging Plastic bottle
Potassium (K) 25ppm max.
Sodium (Na) 50ppm max.
Solubility Solubility in water: soluble. Other solubilities: soluble in methanol, practically insoluble in ethanol, acetone and, ethyl acetate
Strontium (Sr) 1000ppm max.
Trace Analysis 1000ppm max.
Boiling Point 1560.0°C
Color Gray-White
Melting Point 962.0°C
pH 5.2 to 8.2 (5% soln. at 25°C)
Quantity 1kg
Chemical Name or Material Barium chloride dihydrate
Show More Show Less

GHS H Statement
Toxic if swallowed.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear eye protection/face protection.
If eye irritation per

GHS Signal Word: Danger