Learn More
Barium chloride dihydrate, ACS, Thermo Scientific Chemicals
Description
Barium chloride is usually employed as a chemical reagent to precipitate soluble sulfates and also in the purification of the seawater solution. It is practiced in several applications like heat-treating baths, waste water treatment, the production of PVC stabilizers, oil lubricants, fireworks materials, hardening of steel, in the manufacture of pigments and in the manufacture of other barium salts. It is applied to prepare composites to absorb ammonia and to increase the crystallinty of polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 113°C-2H2O |
Physical Form | Crystalline |
Quantity | 500 g |
UN Number | UN1564 |
Merck Index | 14,971 |
Solubility Information | Very soluble in water; Soluble in methanol; Insoluble in ethanol,acetone,ethyl acetate. |
IUPAC Name | barium(2+) dihydrate dichloride |
Formula Weight | 244.28 (208.25 Anhydrous) |
Grade | ACS |
Density | 3.097 |
Show More |
Safety and Handling
Hazard Category | H301-H319-H332 |
Hazard Statement | GHS H Statement H301-H332 |
Precautionary Statement | P261-P264b-P270-P271-P280i-P301+P310-P304+P340-P305+P351+P338-P312-P330-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S. |
EINECSNumber | 233-788-1 |
RTECSNumber | CQ8751000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.