Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Chloride, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
2.5kg
Packaging:
Poly Bottle
10326-27-9
BaCl2H4O2
244.257
PWHCIQQGOQTFAE-UHFFFAOYSA-L
5284346
CHEBI:86153
barium(2+);dichloride;dihydrate
O.O.[Cl-].[Cl-].[Ba+2]
10326-27-9
BaCl2H4O2
244.257
PWHCIQQGOQTFAE-UHFFFAOYSA-L
5284346
CHEBI:86153
barium(2+);dichloride;dihydrate
O.O.[Cl-].[Cl-].[Ba+2]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 244.27
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 3.86kg
Quantity 2.5kg
Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material Barium Chloride, Dihydrate