Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Barium Chloride, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Click to view available options
Quantity:
125g
500g
2.5kg
Packaging:
Poly Bottle
Specifications
Formula Weight | 244.27 |
Physical Form | Crystals |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 3.86kg |
Quantity | 125g |
Chemical Name or Material | Barium Chloride, Dihydrate |