Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Chloride, Dihydrate, Crystal, For Sulfate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

10326-27-9
BaCl2H4O2
244.257
PWHCIQQGOQTFAE-UHFFFAOYSA-L
5284346
CHEBI:86153

High quality chemicals for laboratory and specialized industrial use

    For Sulfate by Turbidimetric Method
CAS 10326-27-9
Molecular Formula BaCl2H4O2
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
SMILES O.O.[Cl-].[Cl-].[Ba+2]
IUPAC Name barium(2+);dichloride;dihydrate
Molecular Weight (g/mol) 244.257
PubChem CID 5284346
ChEBI CHEBI:86153
Formula Weight 244.26
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 3.86kg
Quantity 500g
Chemical Name or Material Barium Chloride, Dihydrate
Grade Baker Analyzed™ ACS Reagent
Show More Show Less