Barium chloride, White beads, 99.99% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1368606

Quantity 5g

 View more versions of this product

Catalog No. AA1368606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10361-37-2
    Physical Form Beads
    Assay Percent Range 99.99% (Metals basis)
    Molecular Formula BaCl2
    Formula Weight 208.25
    MDL Number MFCD00003445
    Chemical Name or Material Barium chloride

    Specifications

    Physical Form Beads
    Formula Weight 208.25
    Quantity 5g
    Color White
    DOT Information DOT Class: 6.1, Packaging Group: III
    EINECS Number 233-788-1
    Odor Odorless
    Assay Percent Range 99.99% (Metals basis)
    Molecular Formula BaCl2
    MDL Number MFCD00003445
    CAS 10361-37-2
    Health Hazard 1 H301-H331: Toxic if swallowed. Toxic if inhaled.
    Chemical Name or Material Barium chloride

    H301-H331: Toxic if swallowed. Toxic if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit