Alfa Aesar™ Barium cyclohexanebutyrate, AAS, Ba 28.9%

Manufacturer: Alfa Aesar™ 8725703

Quantity 1g

 View more versions of this product

Catalog No. AA8725703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 62669-65-2
    Molecular Formula C20H34BaO4
    Formula Weight 475.84
    MDL Number MFCD00036397
    UN Number UN1564
    Synonym Cyclohexanebutyric acid barium salt; 4-Cyclohexylbutyric acid barium salt
    Chemical Name or Material Barium cyclohexanebutyrate
    Grade AAS Grade

    Specifications

    Formula Weight 475.84
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    UN Number UN1564
    Health Hazard 1 Warning
    MDL Number MFCD00036397
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
    Physical Form Powder
    Molecular Formula C20H34BaO4
    CAS 62669-65-2
    Melting Point ≈225°C (dec.)
    Concentration Ba 28.9%
    Synonym Cyclohexanebutyric acid barium salt; 4-Cyclohexylbutyric acid barium salt
    Chemical Name or Material Barium cyclohexanebutyrate
    Grade AAS Grade

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit