Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Diphenylaminesulfonate, 0.1% (w/v) Aqueous Solution, For Residual Chlorine, APHA, Spectrum™

Click to view available options
Quantity:
125 mL
1 L
Packaging:
Poly Bottle
6211-24-1,7732-18-5
6211-24-1,7732-18-5
Grade Chemical Solution
Packaging Poly Bottle
Quantity 1 L

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Material Safety Data Sheet (MSDS) prior to use.