Alfa Aesar™ Barium dodecairon nonadecaoxide

Manufacturer: Alfa Aesar™ 4555222

Quantity 100g

 View more versions of this product

Catalog No. AA4555222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12047-11-9
    Molecular Formula BaFe12O19
    Formula Weight 1111.48
    MDL Number MFCD00075637
    UN Number UN1564
    Synonym Barium ferrite
    Chemical Name or Material Barium dodecairon nonadecaoxide

    Specifications

    Formula Weight 1111.48
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 12047-11-9
    Physical Form Powder
    MDL Number MFCD00075637
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
    Molecular Formula BaFe12O19
    UN Number UN1564
    Color Black
    Melting Point >1315°C
    Synonym Barium ferrite
    Quantity 100g
    Chemical Name or Material Barium dodecairon nonadecaoxide

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit