Alfa Aesar™ Barium fluoride, Puratronic™, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 1064722

Quantity 100g

 View more versions of this product

Catalog No. AA1064722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7787-32-8
    Formula Weight 175.36
    MDL Number MFCD00003450
    UN Number UN1564
    Merck Index 14,974
    Chemical Name or Material Barium fluoride
    Grade Puratronic™

    Specifications

    Formula Weight 175.36
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN1564
    Boiling Point 2260°C
    TSCA TSCA
    Refractive Index 1.4741
    CAS 7787-32-8
    Merck Index 14,974
    Melting Point 1353°C
    Quantity 100g
    Health Hazard 1 Warning
    Density 4.83g/mL
    Odor Odorless
    Physical Form Powder
    MDL Number MFCD00003450
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    Chemical Name or Material Barium fluoride
    Assay 99.99%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit