Alfa Aesar™ Barium hexacyanoferrate(II)

Manufacturer: Alfa Aesar™ A10376-A1

Quantity 1kg

 View more versions of this product

Catalog No. AAA10376A1

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 13821-06-2
    Molecular Formula Ba2Fe(CN)6
    Formula Weight 486.62
    MDL Number MFCD00168098
    UN Number UN1564
    Synonym Barium ferrocyanide
    Chemical Name or Material Barium hexacyanoferrate (II)

    Specifications

    Formula Weight 486.62
    Molecular Formula Ba2Fe(CN)6
    UN Number UN1564
    Health Hazard 2 GHS H Statement
    H331-H302
    Toxic if inhaled.
    Harmful if swallowed.
    CAS 13821-06-2
    Synonym Barium ferrocyanide
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
    MDL Number MFCD00168098
    Quantity 1kg
    Health Hazard 3 GHS P Statement
    P261-P304+P340-P311-P301+P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physic
    Chemical Name or Material Barium hexacyanoferrate (II)

    GHS H Statement
    H331-H302
    Toxic if inhaled.
    Harmful if swallowed.

    GHS P Statement
    P261-P304+P340-P311-P301+P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physic

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit