Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Hydroxide, 8-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

12230-71-6
BaH18O10
315.461
ZUDYPQRUOYEARG-UHFFFAOYSA-L
17749109

High quality chemicals for laboratory and specialized industrial use

SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba+2]
IUPAC Name barium(2+);dihydroxide;octahydrate
Formula Weight 315.46
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.18kg
Quantity 125g
Chemical Name or Material Barium Hydroxide, 8-Hydrate
Grade Baker Analyzed™ ACS Reagent