Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Hydroxide, 8-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
2.5kg
Packaging:
Poly Bottle
12230-71-6
BaH18O10
315.461
ZUDYPQRUOYEARG-UHFFFAOYSA-L
barium hydroxide octahydrate,unii-l5q5v03tbn,l5q5v03tbn,dihydroxybarium octahydrate,barium dihydroxide octahydrate,barium hydroxide octahydrate mi,barium hydroxide ba oh 2 , octahydrate,acmc-1an7p,barium hydroxide octa hydrate,barium hydroxide octa-hydrate
17749109
barium(2+);dihydroxide;octahydrate
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba+2]
12230-71-6
BaH18O10
315.461
ZUDYPQRUOYEARG-UHFFFAOYSA-L
barium hydroxide octahydrate,unii-l5q5v03tbn,l5q5v03tbn,dihydroxybarium octahydrate,barium dihydroxide octahydrate,barium hydroxide octahydrate mi,barium hydroxide ba oh 2 , octahydrate,acmc-1an7p,barium hydroxide octa hydrate,barium hydroxide octa-hydrate
17749109
barium(2+);dihydroxide;octahydrate
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba+2]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 315.46
Physical Form Crystals
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.18kg
Quantity 2.5kg
Chemical Name or Material Barium Hydroxide, 8-Hydrate