Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Hydroxide, Saturated Solution, Spectrum™

Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
Grade Chemical Solution
CAS 12230-71-6
Packaging Poly Bottle
Quantity 1 L