Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium ICP Standard, 1mL = 1mg Ba (1,000ppm Ba), Ricca Chemical

Click to view available options
Quantity:
100mL
500mL
Packaging:
Poly natural
10022-31-8,7697-37-2,7732-18-5
BaN2O6
261.335
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
24798
barium(2+);dinitrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ba+2]
10022-31-8,7697-37-2,7732-18-5
BaN2O6
261.335
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
24798
barium(2+);dinitrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ba+2]
CAS Min % 94.0,4.22,0.18
CAS Max % 97.0,4.39,0.19
Physical Form Liquid
Packaging Poly natural
Concentration or Composition Notes 1,000ppm Ba
Equivalence 1 mL = 1 mg Ba
Color Colorless
Quantity 100mL
Chemical Name or Material Barium ICP Standard