Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Barium iodate monohydrate

Manufacturer:  Alfa Aesar™ 3912918

Quantity 50g

 View more versions of this product

Catalog No. AA3912918


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7787-34-0
505.15 (487.11 Anhydrous)
UN3087
Barium iodate monohydrate
Ba(IO3)2·H2O
MFCD00239354
14,979

Specifications

7787-34-0
Ba(IO3)2·H2O
MFCD00239354
14,979
4.998g/mL
50g
Powder
505.15 (487.11 Anhydrous)
UN3087
Insoluble in alcohol; Soluble in water, nitric acid, hydrochloric acid
Weak
Barium iodate monohydrate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H272-H302-H332
May intensify fire
oxidizer.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P221-P210-P220-P261-P304+P340-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit