Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Barium iodate monohydrate, 96%

Manufacturer:  Alfa Aesar™ A11479-14

Quantity 25g

 View more versions of this product

Catalog No. AAA1147914

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7787-34-0
Ba(IO3)2·H2O
MFCD00239354
14,979
BaI2O6·H2O
505.15 (487.11 Anhydrous)
UN3087
Barium iodate monohydrate

Specifications

7787-34-0
Ba(IO3)2·H2O
MFCD00239354
14,979
Weak
96%
BaI2O6·H2O
505.15 (487.11 Anhydrous)
UN3087
25g
Barium iodate monohydrate

Safety and Handling

GHS H Statement
H272-H302-H332

GHS P Statement
P221-P210-P220-P261-P280-P264-P304+P340-P301+P312-P312-P501a

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit