Alfa Aesar™ Barium iodate monohydrate

Manufacturer: Alfa Aesar™ 3912909

Quantity 10g

 View more versions of this product

Catalog No. AA3912909

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7787-34-0
    Linear Formula Ba(IO3)2·H2O
    Formula Weight 505.15 (487.11 Anhydrous)
    MDL Number MFCD00239354
    UN Number UN3087
    Merck Index 14,979
    Chemical Name or Material Barium iodate monohydrate

    Specifications

    Formula Weight 505.15 (487.11 Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P261-P304+P340-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00239354
    UN Number UN3087
    Merck Index 14,979
    Quantity 10g
    CAS 7787-34-0
    Health Hazard 1 Danger
    Physical Form Powder
    Solubility Information Insoluble in alcohol; Soluble in water, nitric acid, hydrochloric acid
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
    TSCA TSCA
    Odor Weak
    Linear Formula Ba(IO3)2·H2O
    Health Hazard 2 GHS H Statement
    H272-H302-H332
    May intensify fire
    oxidizer.
    Harmful if swallowed.
    Harmful if inhaled.
    Density 4.998g/mL
    Chemical Name or Material Barium iodate monohydrate

    GHS H Statement
    H272-H302-H332
    May intensify fire
    oxidizer.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P221-P210-P220-P261-P304+P340-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit