Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Barium isopropoxide

Manufacturer:  Alfa Aesar™ 1452306

Quantity 5g

 View more versions of this product

Catalog No. AA1452306

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

24363-37-9
255.52
UN3205
Ba[OCH(CH3)2]2
MFCD00050486
Barium isopropoxide

Specifications

24363-37-9
Ba[OCH(CH3)2]2
MFCD00050486
Moisture Sensitive
≈200°C (dec.)
Barium isopropoxide
Powder
255.52
UN3205
Brown to yellow
5g

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H228-H252-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Self-heating in large quantities
may catch fire.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310-P302+P352
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: III; Proper Shipping Name: ALKALINE EARTH METAL ALCOHOLATES, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit