Alfa Aesar™ Barium isopropoxide, 99.5% (metals basis)

Manufacturer: Alfa Aesar™ 4127106

Quantity 5g

 View more versions of this product

Catalog No. AA4127106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 24363-37-9
    Linear Formula Ba[OCH(CH3)2]2
    Formula Weight 255.52
    MDL Number MFCD00050486
    UN Number UN3205
    Chemical Name or Material Barium isopropoxide

    Specifications

    Formula Weight 255.52
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310-P302+P352
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H228-H252-H302-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable solid.
    Self-heating in large quantities
    may catch fire.
    Harmful if swallowed.
    Harmful if inhaled.
    Physical Form Powder/Lump
    Linear Formula Ba[OCH(CH3)2]2
    Sensitivity Moisture Sensitive
    Melting Point ≈200°C (dec.)
    MDL Number MFCD00050486
    CAS 24363-37-9
    UN Number UN3205
    Color White
    Quantity 5g
    DOT Information Transport Hazard Class: 4.2; Packing Group: III; Proper Shipping Name: ALKALINE EARTH METAL ALCOHOLATES, N.O.S.
    Chemical Name or Material Barium isopropoxide
    Assay 99.5%, (metals basis)

    GHS H Statement
    H314-H318-H228-H252-H302-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable solid.
    Self-heating in large quantities
    may catch fire.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310-P302+P352
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit