Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium manganate(VI), 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
7787-35-1
BaMnO4
256.27
MFCD00011514
ZZCNKSMCIZCVDR-UHFFFAOYSA-N
barium manganate, ba.mno4, acmc-1bdpw, manganic acid h2mno4 , barium salt 1:1, barium 2+ ion manganate, barium 2+ ; dioxido dioxo manganese
3084030
barium(2+);dioxido(dioxo)manganese
[O-][Mn](=O)(=O)[O-].[Ba+2]
7787-35-1
BaMnO4
256.27
MFCD00011514
ZZCNKSMCIZCVDR-UHFFFAOYSA-N
barium manganate, ba.mno4, acmc-1bdpw, manganic acid h2mno4 , barium salt 1:1, barium 2+ ion manganate, barium 2+ ; dioxido dioxo manganese
3084030
barium(2+);dioxido(dioxo)manganese
[O-][Mn](=O)(=O)[O-].[Ba+2]
Assay Percent Range 94% min
Linear Formula BaMnO4
Fieser 08,21; 09,23; 10,16; 12,38; 15,18; 16,18
Merck Index 15, 977
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acids
Formula Weight 256.27
Physical Form Fine Crystalline Powder
Percent Purity 95%
Packaging Glass bottle
Color Black-Blue to Blue
Quantity 100g
Chemical Name or Material Barium manganate(VI), 95%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
May intensify fire; oxidizer.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 232-109-6

TSCA : TSCA