Alfa Aesar™ Barium metaborate monohydrate

Manufacturer: Alfa Aesar™ 4582330

Quantity 250g

 View more versions of this product

Catalog No. AA4582330

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 26124-86-7
    Molecular Formula B2BaO4·H2O
    Linear Formula BaB2O4·H2O
    Formula Weight 240.97 (222.96 Anhydrous)
    MDL Number MFCD00046182
    UN Number UN1564
    Synonym Barium borate
    Chemical Name or Material Barium metaborate monohydrate

    Specifications

    Formula Weight 240.97 (222.96 Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline
    Linear Formula BaB2O4·H2O
    CAS 26124-86-7
    Density 3.35g/mL
    TSCA TSCA
    Molecular Formula B2BaO4·H2O
    MDL Number MFCD00046182
    Melting Point >900°C
    Health Hazard 1 Warning
    UN Number UN1564
    Synonym Barium borate
    Color White
    Quantity 250g
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
    Chemical Name or Material Barium metaborate monohydrate

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit