Alfa Aesar™ Barium molybdenum oxide, 99.9% (metals basis excluding Sr)

Electronic and optical equipment, pigment in paints and other protective coatings

Manufacturer: Alfa Aesar™ 4020514

Quantity 25g

 View more versions of this product

Catalog No. AA4020514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7787-37-3
    Formula Weight 297.28
    MDL Number MFCD00014182
    UN Number UN1564
    Synonym Barium molybdate
    Chemical Name or Material Barium molybdenum oxide

    Specifications

    Formula Weight 297.28
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    Mesh Size −100 Mesh
    Odor Odorless
    Physical Form Powder
    Synonym Barium molybdate
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
    UN Number UN1564
    Color White
    Melting Point ≈1600°C
    Solubility Information Slightly soluble in water and acids
    TSCA TSCA
    MDL Number MFCD00014182
    CAS 7787-37-3
    Health Hazard 1 Warning
    Density 4.98g/mL
    Quantity 25g
    Chemical Name or Material Barium molybdenum oxide
    Assay 99.9%, (metals basis excluding Sr)

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit