Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Nitrate, 10% (w/v) Solution, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
10022-31-8
10022-31-8
Grade Chemical Solution
Appearance Clear, Colorless
Packaging Poly Bottle
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.