Learn More
Barium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Description
Barium nitrate is used by the oil and gas industries to make drilling muds. It is used to make paints, bricks, tiles, glass, rubber, and other barium compounds. It also used in is used in signal flares and water softener.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Crystalline |
Quantity | 25 g |
Assay Percent Range | 99.99% (Metal basis) |
IUPAC Name | barium(2+);dinitrate |
Formula Weight | 261.35 |
Appearance | White crystalline |
Density | 3.24g/cm3 |
Chemical Name or Material | Barium nitrate |
Safety and Handling
Hazard Category | H272-H302-H332 |
DOTInformation | DOT Class: 5.1, Packaging Group: II |
EINECSNumber | 233-020-5 |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.