Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium nitrate, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
5g
500g
2.5kg
Packaging:
Glass Bottle
Plastic bottle
10022-31-8
BaN2O6
261.335
MFCD00003442
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
barium nitrate,nitrobarite,nitric acid, barium salt,barium dinitrate,bariumnitrate,dusicnan barnaty czech,nitrato barico spanish,unii-mdc5sw56xc,nitrate de baryum french,barium nitrate ba no3 2
24798
barium(2+);dinitrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ba+2]
10022-31-8
BaN2O6
261.335
MFCD00003442
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
barium nitrate,nitrobarite,nitric acid, barium salt,barium dinitrate,bariumnitrate,dusicnan barnaty czech,nitrato barico spanish,unii-mdc5sw56xc,nitrate de baryum french,barium nitrate ba no3 2
24798
barium(2+);dinitrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ba+2]
Assay Percent Range 99+%
Linear Formula Ba(NO3)2
Merck Index 15, 979
Formula Weight 261.34
Physical Form Crystals
Percent Purity ≥99%
Grade Analysis
Calcium (Ca) 50ppm max.
Chlorine (Cl) 10ppm max.
Density 3.23
Heavy Metals (as Pb) 10ppm max.
Iron (Fe) 5ppm max.
Packaging Glass Bottle
Trace Analysis 1000ppm max.
Color White
pH 5 to 8 (5% soln. at 20°C)
Quantity 5g
Chemical Name or Material Barium nitrate, p.a.
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May intensify fire; oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 233-020-5

RTECSNumber : CQ9625000

TSCA : TSCA