Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium nitrate, ACS reagent, ACROS Organics™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Plastic bottle
10022-31-8
BaN2O6
261.34
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
barium nitrate,nitrobarite,nitric acid, barium salt,barium dinitrate,bariumnitrate,dusicnan barnaty czech,nitrato barico spanish,unii-mdc5sw56xc,nitrate de baryum french,barium nitrate ba no3 2
24798
barium(2+);dinitrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ba+2]
10022-31-8
BaN2O6
261.34
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
barium nitrate,nitrobarite,nitric acid, barium salt,barium dinitrate,bariumnitrate,dusicnan barnaty czech,nitrato barico spanish,unii-mdc5sw56xc,nitrate de baryum french,barium nitrate ba no3 2
24798
barium(2+);dinitrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ba+2]
Linear Formula Ba(NO3)2
Merck Index 15,979
Solubility Information Solubility in water: 9g/100mL (20°C). Other solubilities: practically insoluble in alcohol and acetone
Formula Weight 261.34
Grade ACS Reagent
Chloride 5ppm max
Insoluble Matter 0.01% max
Packaging Plastic bottle
Solubility Solubility in water: 9 g/100mL (20°C). Other solubilities: practically insoluble in alcohol and acetone
Melting Point 592.0°C
Quantity 500g
Chemical Name or Material Barium nitrate
Show More Show Less

GHS H Statement
Toxic if swallowed.
Harmful if inhaled.
May intensify fire; oxidizer.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfor

GHS Signal Word: Danger

EINECSNumber : 233-020-5