Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Nitrate, Crystal, ACS, 99%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
10022-31-8
10022-31-8
Grade ACS
CAS Max % 1
Percent Purity 0.99
Calcium (Ca) 0.0005
Chloride 5 ppm
Heavy Metals 5 ppm
Insoluble Matter 0.0001
Iron (Fe) 2 ppm
Packaging Fiber Drum
Potassium (K) 5e-05
Sodium (Na) 5e-05
Strontium (Sr) 0.001
pH 5.0 to 8.0
Quantity 50 kg
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.