Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Nitrate TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle

B-285, 10022-31-8

Spectrum™ Barium Nitrate TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
CAS 10022-31-8,0.999
CAS Max % 0.001,0.999
Molecular Formula BaN2O6
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name barium(2+) dinitrate
Molecular Weight (g/mol) 261.34
Packaging Amber Glass Bottle
Quantity 500 mL