Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Barium oxalate, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1300922

Quantity 100g

 View more versions of this product

Catalog No. AA1300922


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

516-02-9
225.36
UN1564
Barium oxalate
BaC2O4
MFCD00054429
14,985
Puratronic™

Specifications

516-02-9
BaC2O4
MFCD00054429
14,985
Odorless
400°C (dec.)
Barium oxalate
Puratronic™
Powder
225.36
UN1564
2.658g/mL
White
100g
99.999%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit