Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium oxide, 90%, extra pure, powder, Acros Organics

Click to view available options
Quantity:
25g
500g
2.5kg
Packaging:
Glass Bottle
Plastic bottle
1304-28-5,513-77-9,1304-29-6
BaO
153.34
QVQLCTNNEUAWMS-UHFFFAOYSA-N
barium oxide,baryta,barium monoxide,calcined baryta,barium protoxide,oxyde de baryum french,barium oxide, obtained by calcining witherite,oxyde de baryum
62392
oxobarium
O=[Ba]
1304-28-5,513-77-9,1304-29-6
BaO
153.34
QVQLCTNNEUAWMS-UHFFFAOYSA-N
barium oxide,baryta,barium monoxide,calcined baryta,barium protoxide,oxyde de baryum french,barium oxide, obtained by calcining witherite,oxyde de baryum
62392
oxobarium
O=[Ba]
Aluminum (Al) 500ppm
Assay Percent Range 90%
Linear Formula BaO
Fieser 01,683; 09,23
Merck Index 15,982
Solubility Information Solubility in water: soluble. Other solubilities: soluble in diluted acids,alcohols,insoluble in acetone,nh3,slowly but considerably soluble in methanol,,ethanol forming barium alcoholate
Barium Carbonate 11% max.
Formula Weight 153.34
Physical Form Powder
Percent Purity 90%
Grade Extra Pure
Insoluble Matter 0.5% max. (in HCl)
Iron (Fe) 170ppm
Packaging Glass Bottle
Particle Size < 13 Micron
Silicon Dioxide 0.67%
Sodium (Na) 320ppm
Solubility Solubility in water: soluble. Other solubilities: soluble in diluted acids, alcohols, insoluble in acetone, nh3, slowly but considerably soluble in methanol,, ethanol forming barium alcoholate
Sulfur (S) 300ppm max.
Trace Analysis Trace analysis are typical values
Boiling Point 2000.0°C
Color Gray to Green-Yellow
Melting Point 1920.0°C
Quantity 25g
Chemical Name or Material Barium oxide
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 215-127-9