Barium oxide, Technical grade 90%, Alfa Aesar™

Manufacturer: Alfa Aesar 1219330

Quantity 250g

 View more versions of this product

Catalog No. AA1219330

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1304-28-5
    Physical Form Powder
    Assay Percent Range 90%
    Molecular Formula BaO
    Formula Weight 153.33
    MDL Number MFCD00003453
    Chemical Name or Material Barium oxide
    Grade Technical Grade

    Specifications

    Formula Weight 153.33
    Quantity 250g
    Color White
    DOT Information DOT Class: 6.1, Packaging Group: III
    EINECS Number 215-127-9
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 90%
    Molecular Formula BaO
    Appearance White powder
    MDL Number MFCD00003453
    CAS 1304-28-5
    Health Hazard 1 H302-H332
    Chemical Name or Material Barium oxide
    Grade Technical Grade

    H302-H332

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit