Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium oxide, Technical grade 90%, Alfa Aesar™

Manufacturer:  Alfa Aesar 12193A1

Quantity 1kg

 View more versions of this product

Catalog No. AA12193A1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1304-28-5
90%
153.33
Barium oxide
Powder
BaO
MFCD00003453
Technical Grade

Specifications

1304-28-5
90%
White powder
MFCD00003453
Odorless
1kg
Technical Grade
Powder
BaO
153.33
5.7g/cm3
White
Barium oxide

Safety and Handling

SDS & Safety Documents

H302-H332

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 215-127-9

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit