Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium perchlorate trihydrate, Reagent Grade, Alfa Aesar™

Click to view available options
Quantity:
250g
1kg
10294-39-0
BaCl2H6O11
390.264
MFCD00149148
NBHDMFAHVJWOHD-UHFFFAOYSA-L
barium perchlorate trihydrate,barium perchlorate trihydrate, reagent grade,ba.2clo4.3h2o,acmc-1bt65,barium perchlorate-water 1/2/3,barium 2+ trihydrate diperchlorate
53249218
barium(2+);diperchlorate;trihydrate
O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2]
10294-39-0
BaCl2H6O11
390.264
MFCD00149148
NBHDMFAHVJWOHD-UHFFFAOYSA-L
barium perchlorate trihydrate,barium perchlorate trihydrate, reagent grade,ba.2clo4.3h2o,acmc-1bt65,barium perchlorate-water 1/2/3,barium 2+ trihydrate diperchlorate
53249218
barium(2+);diperchlorate;trihydrate
O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2]
UN Number UN1447
Merck Index 14,987
Solubility Information Soluble in water, methanol; Slightly soluble in ethanol, ethyl acetate, acetone; Insoluble in ether
Formula Weight 390.29 (336.24 Anhydrous)
Physical Form Crystalline
Grade Reagent
Density 2.74 g/mL
Refractive Index 1.533
Quantity 250g
Chemical Name or Material Barium perchlorate trihydrate

GHS H Statement
H271-H302-H332

P210-P220-P221-P261-P264b-P270-P271-P280-P283-P301+P312-P304+P340-P306+P360-P312-P330-P370+P378q-P371+P380+P375-P501c

H271-H302+H332

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: BARIUM PERCHLORATE, SOLID

EINECSNumber : 236-710-4

TSCA : Yes

Recommended Storage : Ambient temperatures