Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Barium rod, 22mm (0.9 in.) dia., 99+% (metals basis), Sr =0.8%

Click to view available options
Quantity:
100g
500g
1kg
7440-39-3
Ba
137.327
MFCD00134031
DSAJWYNOEDNPEQ-UHFFFAOYSA-N
bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom
5355457
CHEBI:32595
barium
[Ba]
7440-39-3
Ba
137.327
MFCD00134031
DSAJWYNOEDNPEQ-UHFFFAOYSA-N
bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom
5355457
CHEBI:32595
barium
[Ba]

Used as getter, in cathode ray tubes, thermionic emitter tubes and sodium vapor lamps

  • Packaged under argon
Weight ≈7.3cm/100g
Quantity 100g
Concentration Sr ≤0.8%
UN Number UN1400
Odor Odorless
Physical Form Rod, ≈7.3cm/100g,Packaged under argon
Percent Purity ≥99%
Color Gray
Chemical Name or Material Barium rod, 22mm (0.9 in.) dia.
Assay (metals basis)

GHS H Statement
H228-H261
Flammable solid.
In contact with water releases flammable gas.

P223-P231+P232-P280-P301+P310-P331-P335+P334-P370+P378q-P501c

H260-H304

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: BARIUM

EINECSNumber : 231-149-1

RTECSNumber : CQ8370000

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures