Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Barium selenite, 97%

Click to view available options
Quantity:
100g
500g
2500mL
13718-59-7
BaO3Se
264.295
MFCD00003446
VQKDPABSEYUERS-UHFFFAOYSA-L
barium selenite,selenious acid, barium salt 1:1,barium 2+ selenite,bariumselenit,barium 2+ selenit,acmc-20alqt,bariumselenite,ba.so3e,selenious acid barium salt,seleniousacid,bariumsalt 1:1
166893
barium(2+);selenite
[O-][Se](=O)[O-].[Ba+2]
13718-59-7
BaO3Se
264.295
MFCD00003446
VQKDPABSEYUERS-UHFFFAOYSA-L
barium selenite,selenious acid, barium salt 1:1,barium 2+ selenite,bariumselenit,barium 2+ selenit,acmc-20alqt,bariumselenite,ba.so3e,selenious acid barium salt,seleniousacid,bariumsalt 1:1
166893
barium(2+);selenite
[O-][Se](=O)[O-].[Ba+2]
Linear Formula BaSeO3
UN Number UN2630
Formula Weight 264.29
Percent Purity 97%
Quantity 100g
Chemical Name or Material Barium selenite

GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c

H300+H310+H330-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENITES

EINECSNumber : 237-280-0

TSCA : Yes

Recommended Storage : Ambient temperatures