Barium silicon oxide, White powder, Alfa Aesar™

Manufacturer: Alfa Aesar 1512222

Quantity 100g

 View more versions of this product

Catalog No. AA1512222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13255-26-0
    Physical Form Powder
    Molecular Formula BaO and SiO2
    Formula Weight 213.4
    MDL Number MFCD00014186
    Synonym Barium metasilicate
    Chemical Name or Material Barium silicon oxide

    Specifications

    Physical Form Powder
    Formula Weight 213.4
    Quantity 100g
    Color White
    DOT Information DOT Class: 6.1, Packaging Group: III
    Molecular Formula BaO and SiO2
    Synonym Barium metasilicate
    MDL Number MFCD00014186
    CAS 13255-26-0
    Health Hazard 1 H302-H332: Harmful if swallowed. Harmful if inhaled.
    Chemical Name or Material Barium silicon oxide

    H302-H332: Harmful if swallowed. Harmful if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit