Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Sulfate, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

7727-43-7
BaO4S
233.383
TZCXTZWJZNENPQ-UHFFFAOYSA-L
24414

High quality chemicals for laboratory and specialized industrial use

CAS 7727-43-7
Molecular Formula BaO4S
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
SMILES [O-]S(=O)(=O)[O-].[Ba+2]
IUPAC Name barium(2+);sulfate
Molecular Weight (g/mol) 233.383
PubChem CID 24414
Formula Weight 233.39
Physical Form Powder
Packaging Poly Bottle
Specific Gravity 1L = 4.50kg
Quantity 500g
Chemical Name or Material Barium Sulfate
Grade BAKER ANALYZED™ Reagent
Show More Show Less