Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Sulfate, USP, 97.5-100.5%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7727-43-7
BaO4S
233.38
TZCXTZWJZNENPQ-UHFFFAOYSA-L
barium(2+) sulfate
[Ba++].[O-]S([O-])(=O)=O
7727-43-7
BaO4S
233.38
TZCXTZWJZNENPQ-UHFFFAOYSA-L
barium(2+) sulfate
[Ba++].[O-]S([O-])(=O)=O

BA125, 7727-43-7, BaSO4

Spectrum™ Barium Sulfate, USP is a contrast medium for CT scans or fluoroscopic studies of the gastrointestinal tract. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Acid Soluble Substances 0.003
Percent Purity 97.5 to 100.5%
Packaging Poly Pail
pH Range 3.5-10.0
Quantity 2.5 kg