Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Barium tetracyanoplatinate(II) tetrahydrate, Premion™, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 3946306

Quantity 5g

 View more versions of this product

Catalog No. AA3946306


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

13755-32-3
MFCD00014183
14,991
Premion™
508.56 (436.50 Anhydrous)
UN1564
Barium tetracyanoplatinate(II) tetrahydrate

Specifications

13755-32-3
508.56 (436.50 Anhydrous)
UN1564
3.05g/mL
5g
Barium tetracyanoplatinate(II) tetrahydrate
Premion™
Powder
MFCD00014183
14,991
100°C (dec.)
Odorless
99.99%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300
Fatal if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit