Alfa Aesar™ Barium tetracyanoplatinate(II) tetrahydrate, Pt 38%

Radiography (making x-ray screens)

Manufacturer: Alfa Aesar™ 1267906

Quantity 5g

 View more versions of this product

Catalog No. AA1267906

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 13755-32-3
    Formula Weight 508.56 (436.50 Anhydrous)
    MDL Number MFCD00014183
    UN Number UN1564
    Merck Index 14,991
    Synonym Barium platinocyanide; Barium cyanoplatinate(II)
    Chemical Name or Material Barium tetracyanoplatinate(II) tetrahydrate

    Specifications

    Formula Weight 508.56 (436.50 Anhydrous)
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym Barium platinocyanide; Barium cyanoplatinate(II)
    Melting Point 100°C (dec.)
    Quantity 5g
    Odor Odorless
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
    UN Number UN1564
    Merck Index 14,991
    Density 3.05g/mL
    Health Hazard 1 Danger
    Physical Form Powder
    MDL Number MFCD00014183
    CAS 13755-32-3
    Health Hazard 2 GHS H Statement
    H300
    Fatal if swallowed.
    Concentration Pt 38%
    Chemical Name or Material Barium tetracyanoplatinate(II) tetrahydrate

    GHS H Statement
    H300
    Fatal if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit