Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Barium tetracyanoplatinate(II) tetrahydrate, Pt 38%

Radiography (making x-ray screens)

Manufacturer:  Alfa Aesar™ 1267903

Quantity 1g

 View more versions of this product

Catalog No. AA1267903

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

13755-32-3
MFCD00014183
14,991
Barium tetracyanoplatinate(II) tetrahydrate
508.56 (436.50 Anhydrous)
UN1564
Barium platinocyanide; Barium cyanoplatinate(II)

Specifications

13755-32-3
508.56 (436.50 Anhydrous)
UN1564
Barium platinocyanide; Barium cyanoplatinate(II)
100°C (dec.)
1g
Barium tetracyanoplatinate(II) tetrahydrate
Powder
MFCD00014183
14,991
3.05g/mL
Pt 38%
Odorless

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300
Fatal if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit