Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium titanate(IV), 99%, ACROS Organics™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Plastic bottle
12047-27-7
BaO3Ti
MFCD00003447
12047-27-7
BaO3Ti
MFCD00003447
CAS Min % 98.5
CAS Max % 100.0
Additional Information SrO: 0.15% max.
Aluminum Oxide 0.04% max.
Assay Percent Range 99%
Linear Formula BaTiO3
Merck Index 15, 996
Formula Weight 233.24
Physical Form Powder
Percent Purity 99%
Density 6.0800g/mL
Loss on Ignition 0.5% max.
Packaging Plastic bottle
Particle Size 0.03% Max. (+325 Mesh), 0.85 to 1.45 Micron (Horiba, D50)
Silicon Dioxide 0.02% max.
Sodium Oxide 0.03% max.
Solubility Solubility in water: insoluble. Other solubilities: insoluble in alkalies, slightly soluble in diluted acids, soluble in concentrated h2so4 and hf
Specific Gravity 6.08
Color White
Melting Point 1625.0°C
Quantity 2.5kg
Chemical Name or Material Barium titanate(IV), 99%
Show More Show Less

GHS P Statement
Harmful if inhaled.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 234-975-0

RTECSNumber : XR1437333

TSCA : TSCA