Alfa Aesar™ Barium titanium(IV) 2-ethylhexanoate pentaisopropoxide, 99.5+% (metals basis), produces 5% BaTiO3, in isopropanol

Manufacturer: Alfa Aesar™ 43893AE

Quantity 100mL

 View more versions of this product

Catalog No. AA43893AE

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C23H50BaO7Ti
    Formula Weight 623.89
    MDL Number MFCD01310249
    UN Number UN1219
    Synonym Barium titanium(IV) 2-ethylhexano-isopropoxide
    Chemical Name or Material Barium titanium (IV) 2-ethylhexanoate pentaisopropoxide, Produces 5% BaTiO3, in isopropanol

    Specifications

    Formula Weight 623.89
    MDL Number MFCD01310249
    UN Number UN1219
    Synonym Barium titanium(IV) 2-ethylhexano-isopropoxide
    Health Hazard 1 Danger
    Quantity 100mL
    Health Hazard 2 GHS H Statement
    H225-H361-H302-H332-H336
    Highly flammable liquid and vapor.
    Suspected of damaging fertility or the unborn child.
    Harmful if swallowed.
    Harmful if inhaled.
    May cause drowsiness or dizziness.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
    Physical Form Liquid
    Molecular Formula C23H50BaO7Ti
    Sensitivity Moisture Sensitive
    Health Hazard 3 GHS P Statement
    P210-P280h-P302+P352
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/clothing.
    IF ON SKIN: Wash with plenty of soap and water.
    Chemical Name or Material Barium titanium (IV) 2-ethylhexanoate pentaisopropoxide, Produces 5% BaTiO3, in isopropanol
    Assay ≥99.5%, (metals basis)

    GHS H Statement
    H225-H361-H302-H332-H336
    Highly flammable liquid and vapor.
    Suspected of damaging fertility or the unborn child.
    Harmful if swallowed.
    Harmful if inhaled.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P280h-P302+P352
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/clothing.
    IF ON SKIN: Wash with plenty of soap and water.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit