Learn More
Barium titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Description
Barium titanium oxide is used for vacuum deposition. It is also used in electronic ceramics, PTC thermistor, the configuration of the capacitors and as a raw material and intermediate in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1620°C |
Physical Form | Pieces |
Quantity | 25 g |
Merck Index | 14,1000 |
Solubility Information | Insoluble in water. |
Formula Weight | 233.19 |
Percent Purity | 99.9% |
Odor | Odorless |
Density | 5.85 g/mL |
Assay | (metals basis) |
Show More |
Safety and Handling
Hazard Category | H302+H332 |
Hazard Statement | GHS H Statement H302-H332 Harmful if swallowed. Harmful if inhaled. |
Precautionary Statement | P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c |
EINECSNumber | 234-975-0 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.