Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium titanium oxide, polymeric precursor, (Oxide ^= 35 wt%), Thermo Scientific Chemicals

Catalog Number AA3975518
Click to view available options
Quantity:
2 g
10 g
50 g
12047-27-7
BaO3Ti
233.19
MFCD00003447
WNKMTAQXMLAYHX-UHFFFAOYSA-N
barium(2+) oxotitaniumbis(olate)
[Ba++].[O-][Ti]([O-])=O
12047-27-7
BaO3Ti
233.19
MFCD00003447
WNKMTAQXMLAYHX-UHFFFAOYSA-N
barium(2+) oxotitaniumbis(olate)
[Ba++].[O-][Ti]([O-])=O

Used in piezoelectric devices

Barium titanium oxide is used in piezoelectric devices. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Viscous Liquid
Quantity 50 g
UN Number UN1993
Solubility Information Soluble in alcohols
Concentration or Composition (by Analyte or Components) Oxide ≈35 wt%
Chemical Name or Material Barium titanium oxide, Polymeric precursor
Hazard Category H302+H332
Hazard Statement GHS H Statement
H226-H302-H332
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA Yes

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit